Learn More
Medchemexpress LLC 4-Nitrophenyl palmitate | 1492-30-4 | 98.0% | 377.52 | 1 G
SDP
Catalog No. 5000454761
Encompass_Preferred
4-Nitrophenyl palmitate is a chromogenic substrate for lipases and esterases. Upon enzymatic hydrolysis, it releases p-nitrophenol, which can be quantified by colorimetric detection at 410 nm as a measure of enzymatic activity. It is used to characterize the activity of various bacterial and mammalian enzymes.
  • Chromogenic substrate for lipases and esterases.
  • Releases p-nitrophenol upon enzymatic hydrolysis.
  • Quantifiable by colorimetric detection at 410 nm.
  • Used to characterize the activity of various bacterial and mammalian enzymes.
  • Serves as a substrate for measuring esterolytic activity of porcine pancreatic lipase and Staphylococcus aureus protease.
Catalog No. 50-004-54761 Supplier Medchemexpress LLC Supplier No. HYW0131681G
4-Nitrophenyl palmitate is a chromogenic substrate for lipases and esterases. Upon enzymatic hydrolysis, it releases p-nitrophenol, which can be quantified by colorimetric detection at 410 nm as a measure of enzymatic activity. It is used to characterize the activity of various bacterial and mammalian enzymes.
  • Chromogenic substrate for lipases and esterases.
  • Releases p-nitrophenol upon enzymatic hydrolysis.
  • Quantifiable by colorimetric detection at 410 nm.
  • Used to characterize the activity of various bacterial and mammalian enzymes.
  • Serves as a substrate for measuring esterolytic activity of porcine pancreatic lipase and Staphylococcus aureus protease.