Learn More
Cayman Chemical 4-Nitrophenyl Palmitate, Purity: ≥95%, CAS: 1492-30-4, 1 g
SDP
Catalog No. 502990396
Encompass_Preferred
A colorimetric lipase and esterase substrate; upon enzymatic hydrolysis, 4-nitrophenol is released which can be quantified by colorimetric detection at 410 nm as a measure of enzyme activity
Catalog No. 50-299-0396 Supplier Cayman Chemical Supplier No. 308701 G
A colorimetric lipase and esterase substrate; upon enzymatic hydrolysis, 4-nitrophenol is released which can be quantified by colorimetric detection at 410 nm as a measure of enzyme activity