Learn More
Cayman Chemical 4-Nitrophenyl Palmitate, Purity: ≥95%, CAS: 1492-30-4, 5 g
SDP
Catalog No. 502989903
Encompass_Preferred
A colorimetric lipase and esterase substrate; upon enzymatic hydrolysis, 4-nitrophenol is released which can be quantified by colorimetric detection at 410 nm as a measure of enzyme activity
Catalog No. 50-298-9903 Supplier Cayman Chemical Supplier No. 308705 G
A colorimetric lipase and esterase substrate; upon enzymatic hydrolysis, 4-nitrophenol is released which can be quantified by colorimetric detection at 410 nm as a measure of enzyme activity