Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(4-Nitrophenyl)(phenyl)iodonium Trifluoromethanesulfonate 98.0+%, TCI America™
SDP

Catalog No. N10665G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N10665G 5 g
N10661G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N10665G Supplier TCI America Supplier No. N10665G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 905718-45-8
Molecular Formula C13H9F3INO5S
Molecular Weight (g/mol) 475.18
MDL Number MFCD21608480
InChI Key OPSOSRFTIUMCJF-UHFFFAOYSA-M
Synonym (4-Nitrophenyl)(phenyl)iodonium Triflate
PubChem CID 16727017
IUPAC Name (4-nitrophenyl)(phenyl)iodanium trifluoromethanesulfonate
SMILES [O-]S(=O)(=O)C(F)(F)F.[O-][N+](=O)C1=CC=C([I+]C2=CC=CC=C2)C=C1

Specifications

Melting Point 200°C
Quantity 5 g
Formula Weight 475.18
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (4-Nitrophenyl)(phenyl)iodonium Trifluoromethanesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.