Learn More
Medchemexpress LLC 4-Nitrophenyl phosph 10g | 333338-18-4 | 371.14 g/mol | C6H4NNa2O6P·6H2O | 10 G
SDP
Catalog No. 5000226742
Encompass_Preferred
4-Nitrophenyl phosphate disodium hexahydrate is a colorless chromogenic substrate used to measure phosphatase activity. Upon enzymatic hydrolysis it yields the yellow 4-nitrophenolate ion, which is commonly monitored by absorbance at 405 nm and used in alkaline phosphatase and protein tyrosine phosphatase assays.
  • Chromogenic substrate for phosphatase activity assays.
  • Produces 4-nitrophenolate with absorbance at 405 nm for spectrophotometric detection.
  • Disodium hexahydrate form improves solubility in aqueous assay buffers.
  • Available in defined pack sizes for assay preparation.
  • High purity suitable for biochemical applications.
Catalog No. 50-002-26742 Supplier Medchemexpress LLC Supplier No. HY116022A10G
4-Nitrophenyl phosphate disodium hexahydrate is a colorless chromogenic substrate used to measure phosphatase activity. Upon enzymatic hydrolysis it yields the yellow 4-nitrophenolate ion, which is commonly monitored by absorbance at 405 nm and used in alkaline phosphatase and protein tyrosine phosphatase assays.
  • Chromogenic substrate for phosphatase activity assays.
  • Produces 4-nitrophenolate with absorbance at 405 nm for spectrophotometric detection.
  • Disodium hexahydrate form improves solubility in aqueous assay buffers.
  • Available in defined pack sizes for assay preparation.
  • High purity suitable for biochemical applications.