Learn More
Medchemexpress LLC 4-Nitrophenyl phosph 10mg | 330-13-2 | 219.09 g/mol | C6H6NO6P | 10 MG
SDP
Catalog No. 5000229734
Encompass_Preferred
4-Nitrophenyl phosphate (CAS 330-13-2) is an organophosphate chromogenic substrate commonly used to measure phosphatase activity. It is supplied as a white to off-white solid (C6H6NO6P, 219.09 g/mol) and is offered in small milligram pack sizes for biochemical and enzymatic studies.
  • Chromogenic substrate for acid and alkaline phosphatase assays.
  • Generates p-nitrophenol upon hydrolysis, enabling spectrophotometric detection.
  • High purity (99.48%) suitable for analytical and enzymatic applications.
  • Solid form simplifies weighing and long-term storage.
  • Available in small milligram pack sizes for research workflows.
Catalog No. 50-002-29734 Supplier Medchemexpress LLC Supplier No. HY11602210MG
4-Nitrophenyl phosphate (CAS 330-13-2) is an organophosphate chromogenic substrate commonly used to measure phosphatase activity. It is supplied as a white to off-white solid (C6H6NO6P, 219.09 g/mol) and is offered in small milligram pack sizes for biochemical and enzymatic studies.
  • Chromogenic substrate for acid and alkaline phosphatase assays.
  • Generates p-nitrophenol upon hydrolysis, enabling spectrophotometric detection.
  • High purity (99.48%) suitable for analytical and enzymatic applications.
  • Solid form simplifies weighing and long-term storage.
  • Available in small milligram pack sizes for research workflows.