Learn More
Medchemexpress LLC 4-Nitrophenyl phosph 1g | 333338-18-4 | 371.14 g/mol | C6H4NNa2O6P·6H2O | 1 G
SDP
Catalog No. 5000229731
Encompass_Preferred
4-Nitrophenyl phosphate disodium hexahydrate is a chromogenic substrate used to measure phosphatase activity; enzymatic hydrolysis yields p-nitrophenol, producing a yellow product that is quantified spectrophotometrically. The material is supplied as an off-white to light yellow solid and is suitable for biochemical assays, ELISA, and general enzyme activity measurements.
  • Chromogenic substrate for alkaline and other phosphatases.
  • Generates p-nitrophenol for spectrophotometric detection.
  • High assay purity for reproducible quantitative results.
  • Supplied as a stable solid for convenient storage and handling.
  • Available in small laboratory sizes for bench-scale experiments.
Catalog No. 50-002-29731 Supplier Medchemexpress LLC Supplier No. HY116022A1G
4-Nitrophenyl phosphate disodium hexahydrate is a chromogenic substrate used to measure phosphatase activity; enzymatic hydrolysis yields p-nitrophenol, producing a yellow product that is quantified spectrophotometrically. The material is supplied as an off-white to light yellow solid and is suitable for biochemical assays, ELISA, and general enzyme activity measurements.
  • Chromogenic substrate for alkaline and other phosphatases.
  • Generates p-nitrophenol for spectrophotometric detection.
  • High assay purity for reproducible quantitative results.
  • Supplied as a stable solid for convenient storage and handling.
  • Available in small laboratory sizes for bench-scale experiments.