Learn More
Medchemexpress LLC 4-Nitrophenyl phosph 5g | 333338-18-4 | 371.14 g/mol | C6H4NO6PNa2·6H2O | 5 G
SDP
Catalog No. 5000226386
Encompass_Preferred
4-Nitrophenyl phosphate disodium hexahydrate is a chromogenic phosphatase substrate that produces yellow 4-nitrophenolate upon enzymatic hydrolysis, enabling absorbance-based activity assays. Supplied as the disodium salt hexahydrate to improve solubility and handling for laboratory use.
  • Sensitive chromogenic substrate for phosphatase activity assays.
  • Generates a measurable signal at 405 nm upon hydrolysis.
  • High purity (≈99.8%) suitable for biochemical assays.
  • Supplied in small laboratory quantities for bench-scale experiments.
  • Stable when stored sealed at -20°C and protected from moisture.
Catalog No. 50-002-26386 Supplier Medchemexpress LLC Supplier No. HY116022A5G
4-Nitrophenyl phosphate disodium hexahydrate is a chromogenic phosphatase substrate that produces yellow 4-nitrophenolate upon enzymatic hydrolysis, enabling absorbance-based activity assays. Supplied as the disodium salt hexahydrate to improve solubility and handling for laboratory use.
  • Sensitive chromogenic substrate for phosphatase activity assays.
  • Generates a measurable signal at 405 nm upon hydrolysis.
  • High purity (≈99.8%) suitable for biochemical assays.
  • Supplied in small laboratory quantities for bench-scale experiments.
  • Stable when stored sealed at -20°C and protected from moisture.