Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-Nitrophenyl phosph 5mg | 330-13-2 | 219.09 g/mol | C6H6NO6P | 5 MG
SDP

Catalog No. 5000229730
Encompass_Preferred

4-nitrophenyl phosphate is a chromogenic phosphate substrate used to measure phosphatase activity. Upon enzymatic hydrolysis it yields the yellow 4-nitrophenolate ion, which is commonly monitored by absorbance at 405 nm. Supplied as a dry powder (5 mg) with high purity; follow recommended storage and handling to maintain activity.

  • Chromogenic substrate for phosphatase assays; produces a yellow product measurable at 405 nm.
  • High purity (99.48%) for reliable analytical and biochemical results.
  • Supplied as a dry powder for easy dissolution and long-term storage.
  • Powder storage: -20°C for long-term stability; aliquot to avoid repeated freeze-thaw cycles.
  • Prepare aqueous working solutions and filter-sterilize (0.22 μm) before use.

Catalog No. 50-002-29730 Supplier Medchemexpress LLC Supplier No. HY1160225MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-nitrophenyl phosphate is a chromogenic phosphate substrate used to measure phosphatase activity. Upon enzymatic hydrolysis it yields the yellow 4-nitrophenolate ion, which is commonly monitored by absorbance at 405 nm. Supplied as a dry powder (5 mg) with high purity; follow recommended storage and handling to maintain activity.

  • Chromogenic substrate for phosphatase assays; produces a yellow product measurable at 405 nm.
  • High purity (99.48%) for reliable analytical and biochemical results.
  • Supplied as a dry powder for easy dissolution and long-term storage.
  • Powder storage: -20°C for long-term stability; aliquot to avoid repeated freeze-thaw cycles.
  • Prepare aqueous working solutions and filter-sterilize (0.22 μm) before use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.