Learn More
Alkali Scientific 4 Nitrophenyl Phosphate bis Cyclohexylammonium Salt, 1 G
SDP
Catalog No. 5000660506
Encompass_Preferred
Nitrophenyl Phosphate bis Cyclohexylammonium Salt, 1g is a reagent commonly used in biochemical applications, especially in enzyme assays and molecular biology experiments. This compound is a versatile substrate for phosphatases and other enzymes that hydrolyze phosphoric esters. The cyclohexylammonium salt form helps improve solubility in various solvents, making it suitable for use in diverse reaction systems. It is used to detect phosphatase activity in biological samples and can be applied in studies involving enzyme kinetics, protein assays, and diagnostic procedures. The reagent is available in a 1g format, making it convenient for laboratory-scale applications.
Catalog No. 50-006-60506 Supplier Alkali Scientific Supplier No. TEB001CP
Nitrophenyl Phosphate bis Cyclohexylammonium Salt, 1g is a reagent commonly used in biochemical applications, especially in enzyme assays and molecular biology experiments. This compound is a versatile substrate for phosphatases and other enzymes that hydrolyze phosphoric esters. The cyclohexylammonium salt form helps improve solubility in various solvents, making it suitable for use in diverse reaction systems. It is used to detect phosphatase activity in biological samples and can be applied in studies involving enzyme kinetics, protein assays, and diagnostic procedures. The reagent is available in a 1g format, making it convenient for laboratory-scale applications.