Learn More
Alkali Scientific 4 Nitrophenyl Phosphate bis Cyclohexylammonium Salt, 25 G
SDP
Catalog No. 5000660507
Encompass_Preferred
Nitrophenyl Phosphate bis Cyclohexylammonium Salt, 25g is a versatile reagent used in biochemical and molecular biology applications. This compound acts as a substrate for various phosphatases and is commonly utilized in enzyme activity assays. The cyclohexylammonium salt enhances the solubility of the compound in different solvents, making it suitable for a broad range of experimental setups. It plays a crucial role in phosphatase assays, protein assays, and enzyme activity studies. The 25g package is ideal for large-scale experiments or for laboratories requiring a bulk supply of the reagent for ongoing applications.
Catalog No. 50-006-60507 Supplier Alkali Scientific Supplier No. TEB002CP
Nitrophenyl Phosphate bis Cyclohexylammonium Salt, 25g is a versatile reagent used in biochemical and molecular biology applications. This compound acts as a substrate for various phosphatases and is commonly utilized in enzyme activity assays. The cyclohexylammonium salt enhances the solubility of the compound in different solvents, making it suitable for a broad range of experimental setups. It plays a crucial role in phosphatase assays, protein assays, and enzyme activity studies. The 25g package is ideal for large-scale experiments or for laboratories requiring a bulk supply of the reagent for ongoing applications.