Learn More
Alkali Scientific 4 Nitrophenyl Phosphate bis Cyclohexylammonium Salt, 5 G
SDP
Catalog No. 5000661017
Encompass_Preferred
Nitrophenyl Phosphate bis Cyclohexylammonium Salt, 5g is used as a reagent in biochemical experiments, particularly in assays involving phosphatases and other enzymes that hydrolyze phosphoric esters. The compound is a reliable substrate for testing enzyme activity, and the cyclohexylammonium salt enhances its solubility in various solutions, making it adaptable to different types of assays. This reagent is ideal for enzyme kinetics studies, protein analysis, and biochemical research. The 5g size is well-suited for smaller experiments or routine laboratory procedures that require a moderate amount of reagent.
Catalog No. 50-006-61017 Supplier Alkali Scientific Supplier No. TEB003CP
Nitrophenyl Phosphate bis Cyclohexylammonium Salt, 5g is used as a reagent in biochemical experiments, particularly in assays involving phosphatases and other enzymes that hydrolyze phosphoric esters. The compound is a reliable substrate for testing enzyme activity, and the cyclohexylammonium salt enhances its solubility in various solutions, making it adaptable to different types of assays. This reagent is ideal for enzyme kinetics studies, protein analysis, and biochemical research. The 5g size is well-suited for smaller experiments or routine laboratory procedures that require a moderate amount of reagent.