Learn More
Alkali Scientific 4 Nitrophenyl Phosphate (Disodium Salt, Hexahydrate), 1 Kg
SDP
Catalog No. 5000661897
Encompass_Preferred
Nitrophenyl Phosphate (Disodium Salt, Hexahydrate) is a chromogenic reagent used for detecting alkaline phosphatase activity in various biological and biochemical assays. The 1kg pack is designed for high-volume laboratories requiring significant quantities of this substrate for enzyme activity measurements. When hydrolyzed by alkaline phosphatase, this compound produces a color change, providing a visual indicator of enzyme activity. This reagent is widely used in clinical diagnostics, microbiological testing, and pharmaceutical research. It is particularly useful for enzyme assays, protein quantification, and enzyme kinetics studies, providing consistent and reproducible results in laboratory settings.
Catalog No. 50-006-61897 Supplier Alkali Scientific Supplier No. ROP001CP
Nitrophenyl Phosphate (Disodium Salt, Hexahydrate) is a chromogenic reagent used for detecting alkaline phosphatase activity in various biological and biochemical assays. The 1kg pack is designed for high-volume laboratories requiring significant quantities of this substrate for enzyme activity measurements. When hydrolyzed by alkaline phosphatase, this compound produces a color change, providing a visual indicator of enzyme activity. This reagent is widely used in clinical diagnostics, microbiological testing, and pharmaceutical research. It is particularly useful for enzyme assays, protein quantification, and enzyme kinetics studies, providing consistent and reproducible results in laboratory settings.