Learn More
Alkali Scientific 4 Nitrophenyl Phosphate Disodium Salt Hexahydrate, 25 G
SDP
Catalog No. 5000661868
Encompass_Preferred
4-Nitrophenyl Phosphate Disodium Salt Hexahydrate, 25g, is an enzyme substrate widely used in biochemical assays to investigate phosphatase activity. The compound is a reliable substrate for various phosphatases, which hydrolyze phosphoric esters, releasing a p-nitrophenol group detectable via spectrophotometry. This disodium salt form is highly soluble in aqueous solutions, making it ideal for a range of biochemical and molecular biology applications. The 25g quantity is perfect for large-scale laboratory experiments or continuous use, especially in enzyme activity studies, protein assays, and other diagnostic applications in biochemistry and pharmacology.
Catalog No. 50-006-61868 Supplier Alkali Scientific Supplier No. ROP004CP
4-Nitrophenyl Phosphate Disodium Salt Hexahydrate, 25g, is an enzyme substrate widely used in biochemical assays to investigate phosphatase activity. The compound is a reliable substrate for various phosphatases, which hydrolyze phosphoric esters, releasing a p-nitrophenol group detectable via spectrophotometry. This disodium salt form is highly soluble in aqueous solutions, making it ideal for a range of biochemical and molecular biology applications. The 25g quantity is perfect for large-scale laboratory experiments or continuous use, especially in enzyme activity studies, protein assays, and other diagnostic applications in biochemistry and pharmacology.