Learn More
Alkali Scientific 4 Nitrophenyl Phosphate Disodium Salt Hexahydrate, 5 G
SDP
Catalog No. 5000661898
Encompass_Preferred
Nitrophenyl Phosphate Disodium Salt Hexahydrate is a biochemical reagent commonly used in various laboratory applications, particularly for enzyme assays and the detection of alkaline phosphatase activity. This reagent contains the disodium salt form of nitrophenyl phosphate, which reacts with phosphatase enzymes to release nitrophenolate, leading to a color change. The hexahydrate form ensures stability and solubility in aqueous solutions. This 5g supply is perfect for small-scale assays and is frequently used in both clinical diagnostics and academic research. It plays a key role in studies involving enzyme kinetics and signal transduction pathways, especially in biochemistry and molecular biology labs.
Catalog No. 50-006-61898 Supplier Alkali Scientific Supplier No. ROP005CP
Nitrophenyl Phosphate Disodium Salt Hexahydrate is a biochemical reagent commonly used in various laboratory applications, particularly for enzyme assays and the detection of alkaline phosphatase activity. This reagent contains the disodium salt form of nitrophenyl phosphate, which reacts with phosphatase enzymes to release nitrophenolate, leading to a color change. The hexahydrate form ensures stability and solubility in aqueous solutions. This 5g supply is perfect for small-scale assays and is frequently used in both clinical diagnostics and academic research. It plays a key role in studies involving enzyme kinetics and signal transduction pathways, especially in biochemistry and molecular biology labs.