Learn More
Alkali Scientific 4 Nitrophenyl Phosphate (Disodium Salt, Hexahydrate), 500 G
SDP
Catalog No. 5000661674
Encompass_Preferred
Nitrophenyl Phosphate (Disodium Salt, Hexahydrate) is a chromogenic substrate commonly used in biochemical assays to measure the activity of alkaline phosphatase. This 500g pack is suitable for large-scale experiments and high-throughput laboratory applications. Upon hydrolysis by alkaline phosphatase, this compound undergoes a color change, which serves as a visual indicator of enzymatic activity. Nitrophenyl Phosphate is used in a variety of diagnostic, research, and pharmaceutical applications, including enzyme kinetics studies and clinical diagnostics. Its high sensitivity and reproducibility make it a valuable tool for monitoring enzyme activity and for conducting routine testing in microbiology and biochemistry.
Catalog No. 50-006-61674 Supplier Alkali Scientific Supplier No. ROP002CP
Nitrophenyl Phosphate (Disodium Salt, Hexahydrate) is a chromogenic substrate commonly used in biochemical assays to measure the activity of alkaline phosphatase. This 500g pack is suitable for large-scale experiments and high-throughput laboratory applications. Upon hydrolysis by alkaline phosphatase, this compound undergoes a color change, which serves as a visual indicator of enzymatic activity. Nitrophenyl Phosphate is used in a variety of diagnostic, research, and pharmaceutical applications, including enzyme kinetics studies and clinical diagnostics. Its high sensitivity and reproducibility make it a valuable tool for monitoring enzyme activity and for conducting routine testing in microbiology and biochemistry.