Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrophenyl Phosphate (Ready-to-use solution) [for ELISA], TCI America™
SDP

Catalog No. N1109100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1109100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1109100ML Supplier TCI America Supplier No. N1109100ML
Add to Cart
Add to Cart

Chemical Identifiers

Synonym p-Nitrophenyl Phosphate, pNPP

Specifications

Color Yellow
Quantity 100 mL
Grade Reagent
Physical Form Liquid
Chemical Name or Material 4-Nitrophenyl Phosphate (Ready-to-use solution) [for ELISA]
TSCA No
Recommended Storage Freezer (-20 °C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.