Learn More
Cayman Chemical 4-Nitrophenyl Phosphate (sodium salt hydrate), Purity: ≥95%, CAS: 333338-18-4, 5 g
SDP
Catalog No. 502949545
Encompass_Preferred
A chromogenic alkaline phosphatase substrate; 4-nitrophenol is released upon hydrolysis by alkaline phosphatase, which can be quantified at 405 nm as a measure of alkaline phosphatase activity
Catalog No. 50-294-9545 Supplier Cayman Chemical Supplier No. 340815 G
A chromogenic alkaline phosphatase substrate; 4-nitrophenol is released upon hydrolysis by alkaline phosphatase, which can be quantified at 405 nm as a measure of alkaline phosphatase activity