Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Nitrophenyl phosphorodichloridate

Catalog No. 1554035G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-540-35G 5 g
15-540-325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15-540-35G Supplier Sigma Aldrich Supplier No. 1554035G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4-Nitrophenyl phosphorodichloridate

Specifications

CAS 777-52-6
Melting Point 44°C to 45°C (lit.)
Boiling Point 182°C to 183°C (13 mmHg)
Molecular Formula C6H4Cl2NO4P
Linear Formula O2NC6H4OP(O)Cl2
MDL Number MFCD00007315
Quantity 5 g
Synonym 4-Nitrophenyl dichlorophosphate
Molecular Weight (g/mol) 255.98
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.