Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrophenylacetic acid, 99%

Catalog No. AC128761000
Change view
Click to view available options
Quantity:
50 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC128761000 100 g
AC128760500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC128761000 Supplier Thermo Scientific Chemicals Supplier No. 128761000
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 104-03-0 | C8H7NO4 | 181.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-03-0
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.15
MDL Number MFCD00007383
InChI Key YBADLXQNJCMBKR-UHFFFAOYSA-N
Synonym 4-nitrophenylacetic acid, 2-4-nitrophenyl acetic acid, p-nitrophenylacetic acid, 4-nitrophenyl acetic acid, 4-nitrobenzeneacetic acid, benzeneacetic acid, 4-nitro, 2-p-nitrophenyl acetic acid, 4-nitrophenylacetate, 4-nitrophenylaceticacid, acetic acid, p-nitrophenyl
PubChem CID 4661
ChEBI CHEBI:40443
SMILES C1=CC(=CC=C1CC(=O)O)[N+](=O)[O-]

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 152°C to 153°C
Color Beige to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Plastic bottle
Linear Formula O2NC6H4CH2CO2H
Quantity 100 g
Beilstein 09, 455
Merck Index 15, 6709
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,chloroform and ether
Formula Weight 181.15
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrophenylacetic acid, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-168-5
RTECSNumber AJ1130010
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.