Learn More
Medchemexpress LLC 4-Nitrophenylboronic acid | 24067-17-2 | MFCD00161360 | 99.95% | C6H6BNO4 | 5 G
SDP
Catalog No. 5000357581
Encompass_Preferred
4-Nitrophenylboronic acid is a biochemical reagent used in life science research. It functions as an arylboronic acid building block for the synthesis of phenols.
  • Biochemical reagent
  • Can be used as a biological material or organic compound
  • Used for life science related research
  • Arylboronic acid building block for synthesizing phenols
Catalog No. 50-003-57581 Supplier Medchemexpress LLC Supplier No. HY508075G
4-Nitrophenylboronic acid is a biochemical reagent used in life science research. It functions as an arylboronic acid building block for the synthesis of phenols.
  • Biochemical reagent
  • Can be used as a biological material or organic compound
  • Used for life science related research
  • Arylboronic acid building block for synthesizing phenols