Learn More
4-Nitrophenylhydrazine mono and dihydrochloride, 98%
Description
4-Nitrophenylhydrazine mono and dihydrochloride exhibits potential anticancer activity. It acts as a derivatizing agent for carbonyl compounds. It is also used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Nitrophenylhydrazine mono and dihydrochloride exhibits potential anticancer activity. It acts as a derivatizing agent for carbonyl compounds. It is also used as pharmaceutical intermediates.
Solubility
Soluble in water, DMSO and methanol.
Notes
Hygroscopic. Incompatible materials are Strong oxidizing agents and moisture. Store under dry inert gas. Protect from humidity nd water.
Specifications
Specifications
| Melting Point | ∽200°C (decomposition) |
| Quantity | 2 g |
| UN Number | UN2811 |
| Sensitivity | Hygroscopic |
| Solubility Information | Soluble in water,DMSO and methanol. |
| Formula Weight | 153.14(f.b.) |
| Percent Purity | 98% |
| Chemical Name or Material | 4-Nitrophenylhydrazine mono and dihydrochloride |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement H331-H302-H312-H315-H319-H335 Toxic if inhaled. Harmful if swallowed. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement P261-P280-P305+P351+P338-P304+P340-P405-P501a Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S. |
| TSCA | Yes |
| Recommended Storage | Store under Nitrogen |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.