Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrophenylketoxime on polystrene, 2% cross-linked, 200-400 mesh, 0.8-1.0 mmol/g

Catalog No. AAL1937603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1937603 1 g
AAL1937606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1937603 Supplier Thermo Scientific Chemicals Supplier No. L1937603
Only null left
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00165078
Synonym Kaiser Oxime resin; 4-Nitrobenzophenone oxime, polymer supported

Specifications

Quantity 1 g
Concentration or Composition (by Analyte or Components) 0.8 to 1.0mmol/g
Mesh Size 200-400 Mesh
Chemical Name or Material 4-Nitrophenylketoxime on polystrene, 2% cross-linked
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.