Learn More
AdipoGen 4-Nitrophthalic anhydride (25 g)
SDP
Supplier:  AdipoGen CDXN0062G025
Encompass_Preferred
Chemical. CAS: 5466-84-2. Formula: C8H3NO5. MW: 193.11. 4-Nitrophthalic anhydride is used an an intermediate for the synthesis of pharmacological agents, polymers and isothiocyanate fluorescent dyes.
Catalog No. 50-205-4551