Learn More
Medchemexpress LLC 4-nitrophthalonitrile | 31643-49-9 | MFCD00040301 | 99.9% | 173.13 g·mol⁻¹ | C8H3N3O2 | 1000 G
SDP
Catalog No. 5000300845
Encompass_Preferred
4-Nitrophthalonitrile is an organic reagent used as a synthetic building block and intermediate for preparing phthalonitrile derivatives and functionalized aromatic compounds. It is supplied as a high-purity, crystalline solid intended for research and development applications in organic synthesis and materials chemistry.
  • High reported purity ≈99.9% (vendor documents show 99.89-99.99%).
  • White to off-white solid suitable for synthetic use.
  • Soluble in DMSO (≈50 mg/mL) with reported protocols for in vivo vehicles.
  • Stable when stored as powder at -20°C; solvent storage conditions specified.
  • Analytical documentation available including COA, SDS, H-NMR, and RP-HPLC.
Catalog No. 50-003-00845 Supplier Medchemexpress LLC Supplier No. HYW0186451000G
4-Nitrophthalonitrile is an organic reagent used as a synthetic building block and intermediate for preparing phthalonitrile derivatives and functionalized aromatic compounds. It is supplied as a high-purity, crystalline solid intended for research and development applications in organic synthesis and materials chemistry.
  • High reported purity ≈99.9% (vendor documents show 99.89-99.99%).
  • White to off-white solid suitable for synthetic use.
  • Soluble in DMSO (≈50 mg/mL) with reported protocols for in vivo vehicles.
  • Stable when stored as powder at -20°C; solvent storage conditions specified.
  • Analytical documentation available including COA, SDS, H-NMR, and RP-HPLC.