Learn More
Accela Chembio Inc 4-nitropicolinic Acid | 1g | 13509-19-8 | MFCD00191562 | 97+% | Shelf Life: 1620 Days | Light Sensitive/moisture Sensitive/nitrogen Or Argon/+4
Supplier:  Accela Chembio Inc SY0050371G
Encompass_Preferred
4-nitropicolinic Acid | 1g | 13509-19-8 | MFCD00191562 | 97+% | Shelf Life: 1620 Days | Light Sensitive/moisture Sensitive/nitrogen Or Argon/+4
Catalog No. 50-354-202