Learn More
Medchemexpress LLC 4-Nitropyrazole | 2075-46-9 | MFCD00159626 | 113.08 | 100 G
SDP
Catalog No. 5000445790
Encompass_Preferred
4-Nitropyrazole is a five-membered nitrogen-containing heterocyclic compound that can be used as a drug intermediate. It can be synthesized into antibiotics/antifungal/antiviral agents containing pyrazole rings. It is also synthesized through C-H activation reactions to produce LRRK2 inhibitors, which are used in the research of Parkinson's disease.
  • Used as a drug intermediate
  • Can be synthesized into antibiotics/antifungal/antiviral agents
  • Used in the research of Parkinson's disease
Catalog No. 50-004-45790 Supplier Medchemexpress LLC Supplier No. HYW002458100G
4-Nitropyrazole is a five-membered nitrogen-containing heterocyclic compound that can be used as a drug intermediate. It can be synthesized into antibiotics/antifungal/antiviral agents containing pyrazole rings. It is also synthesized through C-H activation reactions to produce LRRK2 inhibitors, which are used in the research of Parkinson's disease.
  • Used as a drug intermediate
  • Can be synthesized into antibiotics/antifungal/antiviral agents
  • Used in the research of Parkinson's disease