Learn More
TARGETMOL CHEMICALS INC 4-Nitroquinoline 1-oxide 1G
SDP
Catalog No. 502876449
Encompass_Preferred
Also available in 1 mL, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. 4-Nitroquinoline 1-oxide (4-NQO) is a chemical carcinogen that induces mutations in bacteria, fungi, and animals through the formation of large amounts of purine adducts. Purity 98.06%
Catalog No. 50-287-6449 Supplier TARGETMOL CHEMICALS INC Supplier No. T353661G
Also available in 1 mL, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. 4-Nitroquinoline 1-oxide (4-NQO) is a chemical carcinogen that induces mutations in bacteria, fungi, and animals through the formation of large amounts of purine adducts. Purity 98.06%