Learn More
Selleck Chemical LLC 4-Nitroquinoline 1-oxide
SDP
Supplier:  Selleck Chemical LLC E0155100MG
Encompass_Preferred
4-Nitroquinoline 1-oxide (4-Nitroquinoline-N-oxide 4-NQO 4NQO 4Nqo NQO NQNO) is a highly carcinogenic model chemical that induces mutations in bacteria fungi and animals through the formation of bulky purine adducts 4-Nitroquinoline 1-oxide can be used to induce animal models of Squamous Cell Carcinoma
Catalog No. 50-313-7913