Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Nitroquinoline N-oxide

Catalog No. N8141250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N8141250MG 250 mg
N81411G 1 g
N81415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N8141250MG Supplier Sigma Aldrich Supplier No. N8141250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4-Nitroquinoline N-oxide

Specifications

CAS 56-57-5
Melting Point 154°C to 156°C (lit.)
Molecular Formula C9H6N2O3
Linear Formula C9H6N2O3
MDL Number MFCD00006738
Quantity 250 mg
Synonym 4-NQO; 4-Nitroquinoline 1-oxide
Molecular Weight (g/mol) 190.16
Percent Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.