Learn More
Medchemexpress LLC 4-nitroso hydrochlorothiazide-13C,15N2,d2 | 00-00-0 | MFCD34676448 | 99.3% | 331.73 g·mol⁻1 | C613CH5D2ClN215N2O5S2 | 5 MG
SDP
Catalog No. 5000222577
Encompass_Preferred
4-Nitroso hydrochlorothiazide-13C,15N2,d2 is an isotopically labeled analytical standard intended for use as an internal standard and tracer in mass spectrometry and quantitative assays. The material contains 13C, 15N, and deuterium labels (d2), is supplied as a high-purity solid in research-scale quantities, and is accompanied by manufacturer documentation (SDS and certificate of analysis) for laboratory use.
  • Isotopically labeled with 13C, 15N2, and d2.
  • High purity suitable for quantitative analysis (~99.3%).
  • Suitable as an internal standard for mass spectrometry.
  • Useful for method development and isotopic tracing studies.
  • Supplied in small, research-scale quantities for analytical use.
  • Documentation such as SDS and certificate of analysis available.
Catalog No. 50-002-22577 Supplier Medchemexpress LLC Supplier No. HY144140S5MG
4-Nitroso hydrochlorothiazide-13C,15N2,d2 is an isotopically labeled analytical standard intended for use as an internal standard and tracer in mass spectrometry and quantitative assays. The material contains 13C, 15N, and deuterium labels (d2), is supplied as a high-purity solid in research-scale quantities, and is accompanied by manufacturer documentation (SDS and certificate of analysis) for laboratory use.
  • Isotopically labeled with 13C, 15N2, and d2.
  • High purity suitable for quantitative analysis (~99.3%).
  • Suitable as an internal standard for mass spectrometry.
  • Useful for method development and isotopic tracing studies.
  • Supplied in small, research-scale quantities for analytical use.
  • Documentation such as SDS and certificate of analysis available.