Learn More
Cayman Chemical 4-Nitrosodiphenylamine, Purity: ≥98%, CAS: 156-10-5, 5 g
SDP
Catalog No. 502942519
Encompass_Preferred
A carcinogen; has been found in air samples taken at industrial sites of tire production
Catalog No. 50-294-2519 Supplier Cayman Chemical Supplier No. 407315 G
A carcinogen; has been found in air samples taken at industrial sites of tire production