Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Nitrotoluene

Catalog No. N273221KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N273221KG 1 kg
N27322100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N273221KG Supplier Sigma Aldrich Supplier No. N273221KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4-Nitrotoluene

Specifications

CAS 99-99-0
Melting Point 52°C to 54°C (lit.)
Density 1.392 g/mL (at 25°C (literature))
Boiling Point 238°C (lit.)
Molecular Formula C7H7NO2
Linear Formula CH3C6H4NO2
MDL Number MFCD00007366
Quantity 1 kg
Synonym 1-Methyl-4-nitrobenzene
Molecular Weight (g/mol) 137.14
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.