Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrotoluene, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
1kg
Packaging:
Glass bottle
99-99-0
C7H7NO2
137.14
MFCD00007366
ZPTVNYMJQHSSEA-UHFFFAOYSA-N
4-nitrotoluene,p-nitrotoluene,4-methylnitrobenzene,4-nitrotoluol,benzene, 1-methyl-4-nitro,toluene, p-nitro,p-methylnitrobenzene,para-nitrotoluol,nitrotoluenos,nitrotoluene, p
7473
CHEBI:35227
1-methyl-4-nitrobenzene
CC1=CC=C(C=C1)[N+](=O)[O-]
99-99-0
C7H7NO2
137.14
MFCD00007366
ZPTVNYMJQHSSEA-UHFFFAOYSA-N
4-nitrotoluene,p-nitrotoluene,4-methylnitrobenzene,4-nitrotoluol,benzene, 1-methyl-4-nitro,toluene, p-nitro,p-methylnitrobenzene,para-nitrotoluol,nitrotoluenos,nitrotoluene, p
7473
CHEBI:35227
1-methyl-4-nitrobenzene
CC1=CC=C(C=C1)[N+](=O)[O-]
Packaging Glass bottle
Color Yellow
Quantity 5g
Boiling Point 238.0°C
Infrared Spectrum Authentic
Flash Point 103°C
Melting Point 51.0°C to 54.0°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula CH3C6H4NO2
Beilstein 05, 323
Merck Index 15, 6737
Solubility Information Solubility in water: 0.35g/L (20°C). Other solubilities: soluble in ethanol, ether, benzene, chloroform and, acetone
Formula Weight 137.14
Physical Form Crystalline Solid
Percent Purity 99%
Chemical Name or Material 4-Nitrotoluene, 99%
Show More Show Less

GHS H Statement
May cause damage to organs through prolonged or repeated exposure.
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.
Toxic to aquatic life with long lasting effects.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a po

GHS Signal Word: Danger

EINECSNumber : 202-808-0

RTECSNumber : XT3325000

TSCA : TSCA