Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nonanone, 97+%, Thermo Scientific™

Catalog No. AAA1505014
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1505014 25 g
AAA1505022 100 g
AAA1505036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1505014 Supplier Thermo Scientific Chemicals Supplier No. A1505014

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4485-09-0
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.24
MDL Number MFCD00027286
InChI Key TYBCSQFBSWACAA-UHFFFAOYSA-N
Synonym 4-nonanone, amyl propyl ketone, propyl amyl ketone, pentyl propyl ketone, amylpropylketone, acmc-1ailr, n-pentyl n-propyl ketone
PubChem CID 78236
IUPAC Name nonan-4-one
SMILES CCCCCC(=O)CCC

Specifications

Density 0.819
Boiling Point 187°C
Quantity 25 g
Formula Weight 142.24
Percent Purity ≥97%
Chemical Name or Material 4-Nonanone
EINECSNumber 224-770-4
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.