Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nonanone 98.0+%, TCI America™
SDP

Catalog No. N042725ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N042725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N042725ML Supplier TCI America Supplier No. N042725ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4485-09-0
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.24
MDL Number MFCD00027286
InChI Key TYBCSQFBSWACAA-UHFFFAOYSA-N
Synonym 4-nonanone, amyl propyl ketone, propyl amyl ketone, pentyl propyl ketone, amylpropylketone, acmc-1ailr, n-pentyl n-propyl ketone
PubChem CID 78236
IUPAC Name nonan-4-one
SMILES CCCCCC(=O)CCC

Specifications

Color Yellow
Boiling Point 187°C
Quantity 25 mL
Formula Weight 142.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Nonanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.