Learn More
Cayman Chemical 4-Nonylphenol-2 3 5 6-d4 1mg
SDP
Catalog No. 502916944
Encompass_Preferred
An internal standard for the quantification of 4-nonylphenol by GC- or LC-MS
Catalog No. 50-291-6944 Supplier Cayman Chemical Supplier No. 394971 MG
Add to Cart
Add to Cart
An internal standard for the quantification of 4-nonylphenol by GC- or LC-MS