Learn More
Medchemexpress LLC 4-Nonylphenol-d5 | 358730-95-7 | 99.2% | 1 MG
SDP
Catalog No. 5000341154
Encompass_Preferred
4-Nonylphenol-d5 is the deuterium labeled form of 4-Nonylphenol. This compound is a major degradation product of Nonylphenol ethoxylates (NPEOs), known as a persistent organic pollutant with endocrine-disrupting properties and estrogenic activity.
  • Deuterium labeled compound
  • Can be used as a tracer in various applications
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporation of stable heavy isotopes aids in quantitation during drug development
  • Deuteration may impact pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-41154 Supplier Medchemexpress LLC Supplier No. HY131122S1MG
4-Nonylphenol-d5 is the deuterium labeled form of 4-Nonylphenol. This compound is a major degradation product of Nonylphenol ethoxylates (NPEOs), known as a persistent organic pollutant with endocrine-disrupting properties and estrogenic activity.
  • Deuterium labeled compound
  • Can be used as a tracer in various applications
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporation of stable heavy isotopes aids in quantitation during drug development
  • Deuteration may impact pharmacokinetic and metabolic profiles of drugs