Learn More
Medchemexpress LLC 4-Nonylphenol-d5 | 358730-95-7 | MFCD01861612 | 99.2% | 225.38 g·mol⁻¹ | C15H19D5O | 5 MG
SDP
Catalog No. 5000262569
Encompass_Preferred
4-Nonylphenol-d5 is the deuterium-labeled analog of 4-nonylphenol used as an internal standard or tracer in analytical and environmental studies. It is supplied as a white to off-white solid with high isotopic purity and is suitable for quantification of nonylphenol residues by mass spectrometry-based methods.
  • Deuterium-labeled internal standard for analytical assays.
  • High purity for accurate quantification.
  • Soluble in DMSO at 100 mg/mL; may require ultrasonic agitation.
  • Stable powder storage at -20°C for extended shelf life.
  • White to off-white solid suitable for preparation of standards.
Catalog No. 50-002-62569 Supplier Medchemexpress LLC Supplier No. HY131122S5MG
4-Nonylphenol-d5 is the deuterium-labeled analog of 4-nonylphenol used as an internal standard or tracer in analytical and environmental studies. It is supplied as a white to off-white solid with high isotopic purity and is suitable for quantification of nonylphenol residues by mass spectrometry-based methods.
  • Deuterium-labeled internal standard for analytical assays.
  • High purity for accurate quantification.
  • Soluble in DMSO at 100 mg/mL; may require ultrasonic agitation.
  • Stable powder storage at -20°C for extended shelf life.
  • White to off-white solid suitable for preparation of standards.