Learn More
Medchemexpress LLC 4-Nonylphenol polyethoxylate | 26027-38-3 | 98.0% | 500 MG
SDP
Catalog No. 5000412758
Encompass_Preferred
4-Nonylphenol polyethoxylate (Nonoxynol) is a non-ionic surfactant primarily for research use. It forms degradation products that exhibit estrogenic activity, persist in aquatic environments, and bioaccumulate in the food chain. This product is not sold to patients.
  • Non-ionic surfactant
  • Forms degradation products including 4-nonylphenol
  • Exhibits estrogenic activity
  • Persists in aquatic environments
  • Bioaccumulates in the food chain
Catalog No. 50-004-12758 Supplier Medchemexpress LLC Supplier No. HYB1027500MG
4-Nonylphenol polyethoxylate (Nonoxynol) is a non-ionic surfactant primarily for research use. It forms degradation products that exhibit estrogenic activity, persist in aquatic environments, and bioaccumulate in the food chain. This product is not sold to patients.
  • Non-ionic surfactant
  • Forms degradation products including 4-nonylphenol
  • Exhibits estrogenic activity
  • Persists in aquatic environments
  • Bioaccumulates in the food chain