Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nonyne, 96%, Thermo Scientific™

Catalog No. AAB2134906
Change view
Click to view available options
Quantity:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2134906 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2134906 Supplier Thermo Scientific Chemicals Supplier No. B2134906

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 20184-91-2
Boiling Point 158°C
Flash Point 35°C (95°F)
Molecular Formula C9H16
Linear Formula CH3(CH2)3C≡CCH2CH2CH3
Quantity 5g
MDL Number MFCD00041650
UN Number UN3295
Synonym 4-nonyne, butylpropylacetylene, acmc-209f6g
InChI Key RAYTXPDVKZFUEI-UHFFFAOYSA-N
SMILES CCCCC#CCCC
IUPAC Name non-4-yne
Molecular Weight (g/mol) 124.227
PubChem CID 140650
Formula Weight 124.23
Percent Purity 96%
Chemical Name or Material 4-Nonyne
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
H304-H226
May be fatal if swallowed and enters airways.
Flammable liquid and vapour.
Precautionary Statement GHS P Statement
P210-P280-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.