Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nonyne, TCI America™
SDP

Catalog No. N03785ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N03785ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N03785ML Supplier TCI America Supplier No. N03785ML

Chemical Identifiers

CAS 20184-91-2
Molecular Formula C9H16
Molecular Weight (g/mol) 124.227
MDL Number MFCD00041650
InChI Key RAYTXPDVKZFUEI-UHFFFAOYSA-N
Synonym 4-nonyne, butylpropylacetylene, acmc-209f6g
PubChem CID 140650
IUPAC Name non-4-yne
SMILES CCCCC#CCCC

Specifications

Color Colorless
Boiling Point 158°C
UN Number 3295
Quantity 5 mL
Formula Weight 124.23
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Nonyne
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.