Learn More
Research Products International Corp 4-Nitrophenyl-a-L-fucopyranoside, 100 Milligrams
SDP
Catalog No. 50213530
Encompass_Preferred
Chromogenic substrate for detecting a-L-Fucosidase.
Catalog No. 50-213-530 Supplier Research Products International Corp Supplier No. N120500.1
Chromogenic substrate for detecting a-L-Fucosidase.