Learn More
Research Products International Corp 4-Nitrophenyl-a-L-fucopyranoside, 1 Gram
SDP
Catalog No. 50213531
Encompass_Preferred
Chromogenic substrate for detecting a-L-Fucosidase.
Catalog No. 50-213-531 Supplier Research Products International Corp Supplier No. N120501.0
Chromogenic substrate for detecting a-L-Fucosidase.