Learn More
Cayman Chemical 4-ANPP-d5 (exempt preparation), Purity: ≥99% deuterated forms (d1-d5), CAS: 1189466-15-6, 1 mg
SDP
Catalog No. 502909684
Encompass_Preferred
An internal standard for the quantification of 4-ANPP by GC- or LC-MS; provided as a DEA exempt preparation; intended for research and forensic applications
Catalog No. 50-290-9684 Supplier Cayman Chemical Supplier No. 367811 MG
An internal standard for the quantification of 4-ANPP by GC- or LC-MS; provided as a DEA exempt preparation; intended for research and forensic applications