Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-O-α-D-Glucopyranosyl-D-fructose monohydrate | 207511-09-9 | MFCD00217057 | 99.8% | 360.31 g·mol⁻¹ | C12H24O12 | 50 MG
SDP

Catalog No. 5000281828
Encompass_Preferred

Maltulose monohydrate is a high-purity disaccharide (4-O-α-D-glucopyranosyl-D-fructose monohydrate) supplied as a solid research reagent. It is used as a biomaterial and an energy source for bacteria in life-science applications and includes recommended storage and stability information for laboratory use.

  • High purity (99.82%).
  • Solid form suitable for accurate weighing and formulation.
  • Molecular weight 360.31 g/mol.
  • Recommended storage: -20°C sealed; in solvent -80°C for 6 months.
  • Suitable as an energy source for bacterial cultures and general research use.

Catalog No. 50-002-81828 Supplier Medchemexpress LLC Supplier No. HYW41590950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Maltulose monohydrate is a high-purity disaccharide (4-O-α-D-glucopyranosyl-D-fructose monohydrate) supplied as a solid research reagent. It is used as a biomaterial and an energy source for bacteria in life-science applications and includes recommended storage and stability information for laboratory use.

  • High purity (99.82%).
  • Solid form suitable for accurate weighing and formulation.
  • Molecular weight 360.31 g/mol.
  • Recommended storage: -20°C sealed; in solvent -80°C for 6 months.
  • Suitable as an energy source for bacterial cultures and general research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.