Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-O-β-galactopyranosyl-D-mannopyranose | 20869-27-6 | ≥98.0% | 342.30 g/mol | C12H22O11 | 25 MG
SDP

Catalog No. 5000282903
Encompass_Preferred

4-O-β-Galactopyranosyl-D-mannopyranose is a disaccharide reagent used in glycobiology and carbohydrate research. It has chemical formula C12H22O11, molecular weight 342.30 g/mol, and CAS number 20869-27-6. The compound is supplied in small milligram quantities for laboratory assays; consult the product page and SDS for handling and storage information.

  • Disaccharide reagent for glycobiology applications.
  • Chemical formula C12H22O11 and molecular weight 342.30 g/mol.
  • High purity (≥98.0%) suitable for analytical use.
  • Available in small milligram pack sizes for laboratory assays.
  • CAS number 20869-27-6 for unambiguous identification.

Catalog No. 50-002-82903 Supplier Medchemexpress LLC Supplier No. HYW14549125MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-O-β-Galactopyranosyl-D-mannopyranose is a disaccharide reagent used in glycobiology and carbohydrate research. It has chemical formula C12H22O11, molecular weight 342.30 g/mol, and CAS number 20869-27-6. The compound is supplied in small milligram quantities for laboratory assays; consult the product page and SDS for handling and storage information.

  • Disaccharide reagent for glycobiology applications.
  • Chemical formula C12H22O11 and molecular weight 342.30 g/mol.
  • High purity (≥98.0%) suitable for analytical use.
  • Available in small milligram pack sizes for laboratory assays.
  • CAS number 20869-27-6 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.