Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-O-β-galactopyranosyl-d-mannopyranose | 20869-27-6 | MFCD00070338 | >98.0% | 342.30 g/mol | C12H22O11 | 5 MG
SDP

Catalog No. 5000283041
Encompass_Preferred

4-O-β-galactopyranosyl-d-mannopyranose is a glycosylated disaccharide provided for laboratory research and analytical applications. Supplied in small milligram quantities, the material is intended for use as a biochemical reagent or reference standard; users should consult the certificate of analysis for detailed purity, solubility, and storage instructions.

  • Small-scale research quantity suitable for assay and analytical work.
  • CAS number 20869-27-6 for unambiguous identification.
  • Molecular formula C12H22O11 and molecular weight 342.30 g/mol.
  • High purity, typically ≥98.0% by HPLC.
  • Intended for research use only; not for clinical or human use.
  • Refer to certificate of analysis for storage and handling details.

Catalog No. 50-002-83041 Supplier Medchemexpress LLC Supplier No. HYW1454915MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-O-β-galactopyranosyl-d-mannopyranose is a glycosylated disaccharide provided for laboratory research and analytical applications. Supplied in small milligram quantities, the material is intended for use as a biochemical reagent or reference standard; users should consult the certificate of analysis for detailed purity, solubility, and storage instructions.

  • Small-scale research quantity suitable for assay and analytical work.
  • CAS number 20869-27-6 for unambiguous identification.
  • Molecular formula C12H22O11 and molecular weight 342.30 g/mol.
  • High purity, typically ≥98.0% by HPLC.
  • Intended for research use only; not for clinical or human use.
  • Refer to certificate of analysis for storage and handling details.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.