Learn More
Medchemexpress LLC 4-O-methyl-α-D-glucurono-β-D-xylan (4-O-methylglucuronoxylan) | 9062-57-1 | >95.0% | 25 MG
SDP
Catalog No. 5000283820
Encompass_Preferred
4-O-Methyl-α-D-glucurono-β-D-xylan is a hemicellulosic polysaccharide used as a biochemical reagent and biological material in life-science research. It is commonly applied in studies of plant cell wall structure, xylan characterization, and glycan enzymology. Supplied as a solid, it is intended for research use only.
  • Used for plant cell wall and glycan research.
  • Hemicellulosic polysaccharide suitable for enzymatic and structural studies.
  • Supplied as a solid, off-white to light brown material.
  • Small package sizes available for lab-scale experiments.
  • Storage: powder -20°C for 3 years or 4°C for 2 years; in solvent -80°C for 6 months or -20°C for 1 month.
  • Certificate of analysis available to confirm identity and purity.
Catalog No. 50-002-83820 Supplier Medchemexpress LLC Supplier No. HYW14570125MG
4-O-Methyl-α-D-glucurono-β-D-xylan is a hemicellulosic polysaccharide used as a biochemical reagent and biological material in life-science research. It is commonly applied in studies of plant cell wall structure, xylan characterization, and glycan enzymology. Supplied as a solid, it is intended for research use only.
  • Used for plant cell wall and glycan research.
  • Hemicellulosic polysaccharide suitable for enzymatic and structural studies.
  • Supplied as a solid, off-white to light brown material.
  • Small package sizes available for lab-scale experiments.
  • Storage: powder -20°C for 3 years or 4°C for 2 years; in solvent -80°C for 6 months or -20°C for 1 month.
  • Certificate of analysis available to confirm identity and purity.