Learn More
Medchemexpress LLC HY-U00450 5mg Medchemexpress, 4-O-Methyl honokiol CAS:68592-15-4 Purity:>98%
SDP
Catalog No. 502083197
Encompass_Preferred
Medchemexpress, HY-U00450 5mg 4-O-Methyl honokiol CAS:68592-15-4 4-O-Methyl honokiol is a natural neolignan isolated from Magnolia officinalis, acts as a PPARγ agonist, and inhibtis NF-κB activity, used for cancer and inflammation research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3197 Supplier Medchemexpress LLC Supplier No. HYU004505MG
Medchemexpress, HY-U00450 5mg 4-O-Methyl honokiol CAS:68592-15-4 4-O-Methyl honokiol is a natural neolignan isolated from Magnolia officinalis, acts as a PPARγ agonist, and inhibtis NF-κB activity, used for cancer and inflammation research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.